SY3H5815F951 Synthonix Corporation

3-Methyl-1H-pyrazole-4-carbonitrile

  • Cas Number : 131661-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18620.html

Safety Information

MSDS https://synthonix.com/18620.html
Availability details for
Quantity
  • Availability
  • Add to Cart