SY3H5815F957 Synthonix Corporation

Methyl 3-amino-5-bromo-2-hydroxybenzenecarboxylate

  • Cas Number : 141761-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/19552.html

Safety Information

MSDS https://synthonix.com/19552.html
Availability details for
Quantity
  • Availability
  • Add to Cart