SY3H5815F959 Synthonix Corporation

4-Methylenepiperidine hydrochloride

  • Cas Number : 144230-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/19855.html

Safety Information

MSDS https://synthonix.com/19855.html
Availability details for
Quantity
  • Availability
  • Add to Cart