SY3H5815F95B Synthonix Corporation

2-Methyl-1H-imidazole-5-carboxylic acid

  • Cas Number : 1457-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/19958.html

Safety Information

MSDS https://synthonix.com/19958.html
Availability details for
Quantity
  • Availability
  • Add to Cart