SY3H5815F95D Synthonix Corporation

Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enecarboxylate

  • Cas Number : 151075-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20154.html

Safety Information

MSDS https://synthonix.com/20154.html
Availability details for
Quantity
  • Availability
  • Add to Cart