SY3H5815F966 Synthonix Corporation

5-Methyl-2-(trifluoromethyl)pyridine

  • Cas Number : 1620-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20623.html

Safety Information

MSDS https://synthonix.com/20623.html
Availability details for
Quantity
  • Availability
  • Add to Cart