SY3H5815F96D Synthonix Corporation

(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate hydrochloride

  • Cas Number : 171752-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/21776.html

Safety Information

MSDS https://synthonix.com/21776.html
Availability details for
Quantity
  • Availability
  • Add to Cart