SY3H5815F979 Synthonix Corporation

Methyl 2-Chloro-3-phenylpropionate

  • Cas Number : 18841-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22031.html

Safety Information

MSDS https://synthonix.com/22031.html
Availability details for
Quantity
  • Availability
  • Add to Cart