SY3H5815F97C Synthonix Corporation

6-Methyl-1H-indole-3-carboxylic acid

  • Cas Number : 209920-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/22301.html

Safety Information

MSDS https://synthonix.com/22301.html
Availability details for
Quantity
  • Availability
  • Add to Cart