SY3H5815F97D Synthonix Corporation

Methyl 4-Benzyl-3-morpholinecarboxylate

  • Cas Number : 212650-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22326.html

Safety Information

MSDS https://synthonix.com/22326.html
Availability details for
Quantity
  • Availability
  • Add to Cart