SY3H5815F983 Synthonix Corporation

4-Methoxy-2-methylpyridine

  • Cas Number : 24103-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22575.html

Safety Information

MSDS https://synthonix.com/22575.html
Availability details for
Quantity
  • Availability
  • Add to Cart