SY3H5815F984 Synthonix Corporation

1-Methyl-4-oxocyclohexanecarboxylic acid

  • Cas Number : 24463-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22599.html

Safety Information

MSDS https://synthonix.com/22599.html
Availability details for
Quantity
  • Availability
  • Add to Cart