SY3H5815F990 Synthonix Corporation

Methyl 3-amino-1H-pyrazole-4-carboxylate

  • Cas Number : 29097-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22867.html

Safety Information

MSDS https://synthonix.com/22867.html
Availability details for
Quantity
  • Availability
  • Add to Cart