SY3H5815F993 Synthonix Corporation

1-Methyl-2-oxopyrrolidine-3-carboxylic acid

  • Cas Number : 30932-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22957.html

Safety Information

MSDS https://synthonix.com/22957.html
Availability details for
Quantity
  • Availability
  • Add to Cart