SY3H5815F996 Synthonix Corporation

3-Methyl-5-nitro-1H-pyrazole

  • Cas Number : 34334-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23164.html

Safety Information

MSDS https://synthonix.com/23164.html
Availability details for
Quantity
  • Availability
  • Add to Cart