SY3H5815F9A0 Synthonix Corporation

1-(3-Methyl-1-phenyl-1H-pyrazol-5-yl)piperazine

  • Cas Number : 401566-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23519.html

Safety Information

MSDS https://synthonix.com/23519.html
Availability details for
Quantity
  • Availability
  • Add to Cart