SY3H5815F9BC Synthonix Corporation

Methyl 4-(benzyloxy)-3-methoxybenzoate

  • Cas Number : 56441-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24295.html

Safety Information

MSDS https://synthonix.com/24295.html
Availability details for
Quantity
  • Availability
  • Add to Cart