SY3H5815F9BD Synthonix Corporation

Methyl 2-bromo-4-chlorobenzoate

  • Cas Number : 57381-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24338.html

Safety Information

MSDS https://synthonix.com/24338.html
Availability details for
Quantity
  • Availability
  • Add to Cart