SY3H5815F9D9 Synthonix Corporation

Methyl 5-hydroxy-2-methylbenzoate

  • Cas Number : 73505-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25125.html

Safety Information

MSDS https://synthonix.com/25125.html
Availability details for
Quantity
  • Availability
  • Add to Cart