SY3H5815F9E5 Synthonix Corporation

4-Methylthiophene-3-carboxylic acid

  • MDL number MFCD02257717
  • Cas Number : 78071-30-4
  • Molecular Weight : 142.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/25374.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/25374.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart