SY3H5815F9EA Synthonix Corporation

5-Methyl-1H-indazol-4-amine

  • Cas Number : 81115-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25495.html

Safety Information

MSDS https://synthonix.com/25495.html
Availability details for
Quantity
  • Availability
  • Add to Cart