SY3H5815F9F1 Synthonix Corporation

Methyl 3-bromo-4-fluoro-1H-indazole-6-carboxylate

  • Cas Number : 885521-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26150.html

Safety Information

MSDS https://synthonix.com/26150.html
Availability details for
Quantity
  • Availability
  • Add to Cart