SY3H5815F9F4 Synthonix Corporation

3-Methoxy-5-methylpyrazin-2-amine

  • Cas Number : 89464-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26266.html

Safety Information

MSDS https://synthonix.com/26266.html
Availability details for
Quantity
  • Availability
  • Add to Cart