SY3H5815F9FD Synthonix Corporation

Methyl 1H-pyrrolo[3,2-b]pyridine-3-carboxylate

  • Cas Number : 952800-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26864.html

Safety Information

MSDS https://synthonix.com/26864.html
Availability details for
Quantity
  • Availability
  • Add to Cart