SY3H5815FA56 Synthonix Corporation

5-Methylisoquinolin-1(2H)-one

  • Cas Number : 24188-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/9887.html

Safety Information

MSDS https://synthonix.com/9887.html
Availability details for
Quantity
  • Availability
  • Add to Cart