SY3H5815FA5E Synthonix Corporation

5-Nitro-1H-indazol-6-ol

  • Cas Number : 1082041-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16449.html

Safety Information

MSDS https://synthonix.com/16449.html
Availability details for
Quantity
  • Availability
  • Add to Cart