SY3H5815FA60 Synthonix Corporation

6-Nitro-1,2,3,4-tetrahydroquinoline

  • Cas Number : 14026-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/19437.html

Safety Information

MSDS https://synthonix.com/19437.html
Availability details for
Quantity
  • Availability
  • Add to Cart