SY3H5815FA69 Synthonix Corporation

3-Nitro-4-chlorobenzenesulfonamide

  • Cas Number : 97-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26959.html

Safety Information

MSDS https://synthonix.com/26959.html
Availability details for
Quantity
  • Availability
  • Add to Cart