SY3H5815FA6A Synthonix Corporation

5-Nitro-3-(trifluoromethyl)pyridin-2-ol

  • MDL number MFCD00276983
  • Cas Number : 99368-66-8
  • Molecular Weight : 208.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to light-yellow to yellow-brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/27001.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/27001.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart