SY3H5815FA6C Synthonix Corporation

2-Nitroisophthalic acid

  • MDL number MFCD00017007
  • Cas Number : 21161-11-5
  • Molecular Weight : 211.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Pale yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/28726.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/28726.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart