SY3H5815FA81 Synthonix Corporation

1H-Pyrazolo[4,3-b]pyridine-5-carbonitrile

  • Cas Number : 1033772-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16159.html

Safety Information

MSDS https://synthonix.com/16159.html
Availability details for
Quantity
  • Availability
  • Add to Cart