SY3H5815FA95 Synthonix Corporation

4-(Pyrrolidin-1-yl)piperidine dihydrochloride

  • Cas Number : 4983-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23929.html

Safety Information

MSDS https://synthonix.com/23929.html
Availability details for
Quantity
  • Availability
  • Add to Cart