SY3H5815FA96 Synthonix Corporation

Pyrimidine-4,6-dicarbonitrile

  • Cas Number : 50844-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23998.html

Safety Information

MSDS https://synthonix.com/23998.html
Availability details for
Quantity
  • Availability
  • Add to Cart