SY3H5815FAA4 Synthonix Corporation

1H-Pyrrolo[3,2-c]pyridine-2-carbaldehyde

  • Cas Number : 630395-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24641.html

Safety Information

MSDS https://synthonix.com/24641.html
Availability details for
Quantity
  • Availability
  • Add to Cart