SY3H5815FAA9 Synthonix Corporation

1-Phenyl-1H-pyrazol-5-amine

  • Cas Number : 826-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/25545.html

Safety Information

MSDS https://synthonix.com/25545.html
Availability details for
Quantity
  • Availability
  • Add to Cart