SY3H5815FAB1 Synthonix Corporation

Pyridazine-3,6-diamine

  • Cas Number : 61070-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/3643.html

Safety Information

MSDS https://synthonix.com/3643.html
Availability details for
Quantity
  • Availability
  • Add to Cart