SY3H5815FAB9 Synthonix Corporation

Pentafluorophenylboronic acid

  • Cas Number : 1582-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/4880.html

Safety Information

MSDS https://synthonix.com/4880.html
Availability details for
Quantity
  • Availability
  • Add to Cart