SY3H5815FAC0 Synthonix Corporation

4-Pyrrolidin-2-ylpyridine

  • Cas Number : 128562-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8797.html

Safety Information

MSDS https://synthonix.com/8797.html
Availability details for
Quantity
  • Availability
  • Add to Cart