SY3H5815FADF Synthonix Corporation

Spiro[indoline-3,4'-piperidin]-2-one hydrochloride

  • Cas Number : 356072-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23260.html

Safety Information

MSDS https://synthonix.com/23260.html
Availability details for
Quantity
  • Availability
  • Add to Cart