SY3H5815FAE0 Synthonix Corporation

Spiro[piperidine-4,3'-pyrrolo[2,3-b]pyridin]-2'(1'H)-one

  • Cas Number : 884049-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26069.html

Safety Information

MSDS https://synthonix.com/26069.html
Availability details for
Quantity
  • Availability
  • Add to Cart