SY3H5815FAE9 Synthonix Corporation

(R)-4,5,6,7-Tetrahydro-benzothiazole-2,6-diamine

  • Cas Number : 106092-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16346.html

Safety Information

MSDS https://synthonix.com/16346.html
Availability details for
Quantity
  • Availability
  • Add to Cart