SY3H5815FAEF Synthonix Corporation

(4-(Trifluoromethyl)pyrimidin-2-yl)methanol

  • Cas Number : 1240594-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17924.html

Safety Information

MSDS https://synthonix.com/17924.html
Availability details for
Quantity
  • Availability
  • Add to Cart