SY3H5815FAF0 Synthonix Corporation

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1H)-one

  • Cas Number : 1256358-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18070.html

Safety Information

MSDS https://synthonix.com/18070.html
Availability details for
Quantity
  • Availability
  • Add to Cart