SY3H5815FB07 Synthonix Corporation

2,3,3-Trimethyl-5-nitro-3H-indole

  • Cas Number : 3484-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23207.html

Safety Information

MSDS https://synthonix.com/23207.html
Availability details for
Quantity
  • Availability
  • Add to Cart