SY3H5815FB0C Synthonix Corporation

3-(Trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyrazine

  • Cas Number : 486460-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23867.html

Safety Information

MSDS https://synthonix.com/23867.html
Availability details for
Quantity
  • Availability
  • Add to Cart