SY3H5815FB13 Synthonix Corporation

O-(Tetrahydropyran-2-yl)-hydroxylamine

  • Cas Number : 6723-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24832.html

Safety Information

MSDS https://synthonix.com/24832.html
Availability details for
Quantity
  • Availability
  • Add to Cart