SY3H5815FB15 Synthonix Corporation

3-(Trifluoromethyl)thiophene-2-carboxylic acid

  • Cas Number : 767337-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25306.html

Safety Information

MSDS https://synthonix.com/25306.html
Availability details for
Quantity
  • Availability
  • Add to Cart