SY3H5815FB17 Synthonix Corporation

1,3,7-Triazaspiro[4.5]decane-2,4-dione

  • Cas Number : 78222-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25378.html

Safety Information

MSDS https://synthonix.com/25378.html
Availability details for
Quantity
  • Availability
  • Add to Cart