SY3H5815FB20 Synthonix Corporation

6-(Trifluoromethyl)-1H-indazole

  • Cas Number : 954239-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26875.html

Safety Information

MSDS https://synthonix.com/26875.html
Availability details for
Quantity
  • Availability
  • Add to Cart