SY3H5815FB26 Synthonix Corporation

3-(Thiophen-2-yl)-1H-pyrazole-4-carbaldehyde

  • Cas Number : 26033-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3270.html

Safety Information

MSDS https://synthonix.com/3270.html
Availability details for
Quantity
  • Availability
  • Add to Cart